COM448617850 Combi-Blocks, Inc.

2-Amino-5-cyanobenzamide

  • MDL number MFCD16877169
  • Cas Number : 606490-51-1
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5784
Availability details for
Quantity
  • Availability
  • Add to Cart