COM448617863 Combi-Blocks, Inc.

Neopentyl 2-chloro-2-oxoacetate

  • MDL number MFCD17214254
  • Cas Number : 209848-82-8
  • Molecular Weight : 178.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-0546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-0546
Availability details for
Quantity
  • Availability
  • Add to Cart