COM448617865 Combi-Blocks, Inc.

2-(3,4-Dihydroxyphenyl)-3-hydroxy-6-methylchromen-4-one

  • MDL number MFCD12547403
  • Cas Number : 260063-32-9
  • Molecular Weight : 284.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-0548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-0548
Availability details for
Quantity
  • Availability
  • Add to Cart