COM448617870 Combi-Blocks, Inc.

Ethyl 6-methyl-4-oxo-6H,7H-pyrazolo[3,2-c][1,4]oxazine-2-carboxylate

  • MDL number MFCD28143930
  • Cas Number : 1820649-45-3
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-4235

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CS-4235
Availability details for
Quantity
  • Availability
  • Add to Cart