COM448617876 Combi-Blocks, Inc.

Cinnamylidencyanothioacetamide

  • MDL number MFCD00119186
  • Cas Number : 91974-48-0
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-6115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-6115
Availability details for
Quantity
  • Availability
  • Add to Cart