COM448617880 Combi-Blocks, Inc.

2-Bromo-5-methylsulfonylnitrobenzene

  • MDL number MFCD00054605
  • Cas Number : 94832-06-1
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9011
Availability details for
Quantity
  • Availability
  • Add to Cart