COM448617896 Combi-Blocks, Inc.

N-Thiomorpholinyl 4-bromobenzenesulfonamide

  • MDL number MFCD07783890
  • Cas Number : 223555-81-5
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9094
Availability details for
Quantity
  • Availability
  • Add to Cart