COM448617897 Combi-Blocks, Inc.

Dimethyl 3,3'-bipyridine-5,5'-dicarboxylate

  • MDL number MFCD07363810
  • Cas Number : 128612-43-1
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9096
Availability details for
Quantity
  • Availability
  • Add to Cart