COM448617906 Combi-Blocks, Inc.

2,4,6-Trimethyl-N'-(4-methylcyclohexylidene)benzenesulfonohydrazide

  • MDL number MFCD22383686
  • Cas Number : 1393441-80-9
  • Molecular Weight : 308.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9130
Availability details for
Quantity
  • Availability
  • Add to Cart