COM448617920 Combi-Blocks, Inc.

2,5-Di(2,5,8,11,14-pentaoxapentadecyl)-1,4-phenylenediboronic acid

  • MDL number MFCD22383687
  • Molecular Weight : 606.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9166
Availability details for
Quantity
  • Availability
  • Add to Cart