COM448617922 Combi-Blocks, Inc.

Ethyl 6,7-dichloro-3-trifluoromethyl-1,4-dihydroquinoxaline-2-carboxylate

  • MDL number MFCD09878367
  • Cas Number : 957062-92-9
  • Molecular Weight : 341.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9168
Availability details for
Quantity
  • Availability
  • Add to Cart