COM448617926 Combi-Blocks, Inc.

3-Hydroxy-2-[3-hydroxy-4-(pyrrolidin-1-yl)phenyl]benzo[h]chromen-4-one

  • MDL number MFCD22192466
  • Cas Number : 1353224-63-1
  • Molecular Weight : 373.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9181
Availability details for
Quantity
  • Availability
  • Add to Cart