COM448617928 Combi-Blocks, Inc.

2-[4-(Cyclopentylamino)-3-hydroxyphenyl]-3-hydroxybenzo[h]chromen-4-one

  • MDL number MFCD22192468
  • Cas Number : 1373232-43-9
  • Molecular Weight : 387.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9183
Availability details for
Quantity
  • Availability
  • Add to Cart