COM448617929 Combi-Blocks, Inc.

(2E)-3-[3-Hydroxy-4-(pyrrolidin-1-yl)phenyl]-1-(1-hydroxynaphthalen-2-yl)prop-2-en-1-one

  • MDL number MFCD22574942
  • Cas Number : 1414079-57-4
  • Molecular Weight : 359.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9184
Availability details for
Quantity
  • Availability
  • Add to Cart