COM448617936 Combi-Blocks, Inc.

N,N-Diethyl 2-bromobenzamide

  • MDL number MFCD00457139
  • Cas Number : 76041-86-6
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9401

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9401
Availability details for
Quantity
  • Availability
  • Add to Cart