COM448617937 Combi-Blocks, Inc.

1-(Pyrrolidin-1-ylcarbonyl)-2-bromobenzene

  • MDL number MFCD00590304
  • Cas Number : 124461-27-4
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9402
Availability details for
Quantity
  • Availability
  • Add to Cart