COM448617943 Combi-Blocks, Inc.

N-Butyl 2-bromobenzamide

  • MDL number MFCD01212979
  • Cas Number : 349403-39-0
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CS-9410
Availability details for
Quantity
  • Availability
  • Add to Cart