COM448617962 Combi-Blocks, Inc.

(5-Chloro-2-[(2,2-dimethylpropoxy)carbonyl]phenyl)boronic acid

  • MDL number MFCD01631848
  • Cas Number : 1315476-05-1
  • Molecular Weight : 270.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1031
Availability details for
Quantity
  • Availability
  • Add to Cart