COM448617983 Combi-Blocks, Inc.

1H-Benzimidazol-4-ylboronic acid

  • MDL number MFCD05664677
  • Cas Number : 499769-95-8
  • Molecular Weight : 162.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1113
Availability details for
Quantity
  • Availability
  • Add to Cart