COM448617986 Combi-Blocks, Inc.

5-Methyl-1-hexenylboronic acid

  • MDL number MFCD06202463
  • Cas Number : 197724-98-4
  • Molecular Weight : 142.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1143
Availability details for
Quantity
  • Availability
  • Add to Cart