COM448617989 Combi-Blocks, Inc.

1H-Pyrazole-1-benzyl-4-boronic acid

  • MDL number MFCD06248750
  • Molecular Weight : 202.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1303

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1303
Availability details for
Quantity
  • Availability
  • Add to Cart