COM448618001 Combi-Blocks, Inc.

2-Methoxy-3-methylphenyl boronic acid

  • MDL number MFCD06801731
  • Cas Number : 909187-39-9
  • Molecular Weight : 166.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1341
Availability details for
Quantity
  • Availability
  • Add to Cart