COM448618006 Combi-Blocks, Inc.

4-(2,2-Dimethoxyethoxy)phenylboronic acid

  • MDL number MFCD06801723
  • Cas Number : 1256355-36-8
  • Molecular Weight : 226.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1357

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1357
Availability details for
Quantity
  • Availability
  • Add to Cart