COM448618038 Combi-Blocks, Inc.

3,5-Difluoro-4-difluoromethoxy-benzeneboronic acid

  • MDL number MFCD08272029
  • Cas Number : 915401-97-7
  • Molecular Weight : 223.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1503

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1503
Availability details for
Quantity
  • Availability
  • Add to Cart