COM448618043 Combi-Blocks, Inc.

3'-Trifluoromethylbiphenyl-4-boronic acid

  • MDL number MFCD08544398
  • Cas Number : 1106837-36-8
  • Molecular Weight : 266.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1515
Availability details for
Quantity
  • Availability
  • Add to Cart