COM448618048 Combi-Blocks, Inc.

2-P-Terphenylboronic acid

  • MDL number MFCD08669638
  • Cas Number : 663954-31-2
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1526

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1526
Availability details for
Quantity
  • Availability
  • Add to Cart