COM448618051 Combi-Blocks, Inc.

4-(Cyclohexylsulfanyl)phenylboronic acid

  • MDL number MFCD08701753
  • Cas Number : 1107603-50-8
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1539
Availability details for
Quantity
  • Availability
  • Add to Cart