COM448618054 Combi-Blocks, Inc.

3-Benzyloxy-4-chlorophenylboronic acid

  • MDL number MFCD08701723
  • Cas Number : 1007170-24-2
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1546
Availability details for
Quantity
  • Availability
  • Add to Cart