COM448618055 Combi-Blocks, Inc.

2-Methoxycarbonyl-4-methoxyphenylboronic acid

  • MDL number MFCD08704672
  • Cas Number : 1256355-40-4
  • Molecular Weight : 210.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1547
Availability details for
Quantity
  • Availability
  • Add to Cart