COM448618061 Combi-Blocks, Inc.

4-(3-Methylphenyl)phenylboronic acid

  • MDL number MFCD08701725
  • Cas Number : 501944-56-5
  • Molecular Weight : 212.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1558
Availability details for
Quantity
  • Availability
  • Add to Cart