COM448618063 Combi-Blocks, Inc.

4-(2-methoxyphenyl)phenylboronic acid

  • MDL number MFCD08701729
  • Cas Number : 1107041-07-5
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1561
Availability details for
Quantity
  • Availability
  • Add to Cart