COM448618065 Combi-Blocks, Inc.

4-Butyrylphenylboronic acid

  • MDL number MFCD08701778
  • Cas Number : 186498-24-8
  • Molecular Weight : 192.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1572
Availability details for
Quantity
  • Availability
  • Add to Cart