COM448618066 Combi-Blocks, Inc.

2-(N-Phenylaminomethyl)phenylboronic acid

  • MDL number MFCD08701786
  • Cas Number : 327096-48-0
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1575
Availability details for
Quantity
  • Availability
  • Add to Cart