COM448618069 Combi-Blocks, Inc.

3-Fluoro-2-(trifluoromethyl)phenylboronic acid

  • MDL number MFCD08706086
  • Cas Number : 850411-12-0
  • Molecular Weight : 207.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1581
Availability details for
Quantity
  • Availability
  • Add to Cart