COM448618077 Combi-Blocks, Inc.

3,4-Difluoro-5-trifluoromethylphenylboronic acid

  • MDL number MFCD08706255
  • Cas Number : 864759-64-8
  • Molecular Weight : 225.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1598
Availability details for
Quantity
  • Availability
  • Add to Cart