COM448618091 Combi-Blocks, Inc.

2-(Cyclopentyl)ethenyl-1-boronic acid

  • MDL number MFCD09840294
  • Cas Number : 161282-93-5
  • Molecular Weight : 140.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1671
Availability details for
Quantity
  • Availability
  • Add to Cart