COM448618097 Combi-Blocks, Inc.

5-tert-Butoxycarbonylthiophene-2-boronic acid

  • MDL number MFCD09952055
  • Cas Number : 925921-29-5
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1706
Availability details for
Quantity
  • Availability
  • Add to Cart