COM448618098 Combi-Blocks, Inc.

4-(Phenoxymethyl)phenylboronic acid

  • MDL number MFCD09966155
  • Cas Number : 397843-61-7
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1709

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1709
Availability details for
Quantity
  • Availability
  • Add to Cart