COM448618103 Combi-Blocks, Inc.

4-Borono-N-methylpyridinium iodide

  • MDL number MFCD09952978
  • Cas Number : 362045-65-6
  • Molecular Weight : 264.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1718
Availability details for
Quantity
  • Availability
  • Add to Cart