COM448618104 Combi-Blocks, Inc.

2-Chloro-4-fluoro-5-methoxyphenylboronic acid

  • MDL number MFCD09998845
  • Cas Number : 1256355-46-0
  • Molecular Weight : 204.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-fluoro-5-methoxyphenylboronic acid
Inchi Code 1S/C7H7BClFO3/c1-13-7-2-4(8(11)12)5(9)3-6(7)10/h2-3,11-12H,1H3
Inchi Key KMSUZUNCVLTXJB-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1723
Availability details for
Quantity
  • Availability
  • Add to Cart