COM448618115 Combi-Blocks, Inc.

1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-boronic acid

  • MDL number MFCD10696651
  • Cas Number : 521985-24-0
  • Molecular Weight : 176.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1784

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1784
Availability details for
Quantity
  • Availability
  • Add to Cart