COM448618120 Combi-Blocks, Inc.

3-Chloro-4-(trifluoromethoxy)phenylboronic acid

  • MDL number MFCD10697423
  • Cas Number : 870822-79-0
  • Molecular Weight : 240.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1804

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1804
Availability details for
Quantity
  • Availability
  • Add to Cart