COM448618122 Combi-Blocks, Inc.

(2-Ethynylphenyl)boronic acid

  • MDL number MFCD10697431
  • Cas Number : 905926-85-4
  • Molecular Weight : 146.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1811

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1811
Availability details for
Quantity
  • Availability
  • Add to Cart