COM448618124 Combi-Blocks, Inc.

4-(3-Chlorophenylmethoxy)phenylboronic acid

  • MDL number MFCD10698491
  • Cas Number : 871125-96-1
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1819
Availability details for
Quantity
  • Availability
  • Add to Cart