COM448618127 Combi-Blocks, Inc.

3-Chloro-4,5-difluorophenylboronic acid

  • MDL number MFCD10697406
  • Cas Number : 1643467-84-8
  • Molecular Weight : 192.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1829
Availability details for
Quantity
  • Availability
  • Add to Cart