COM448618136 Combi-Blocks, Inc.

3-Bromo-5-methoxypyridine-4-boronic acid

  • MDL number MFCD11040271
  • Cas Number : 612845-45-1
  • Molecular Weight : 231.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1873
Availability details for
Quantity
  • Availability
  • Add to Cart