COM448618158 Combi-Blocks, Inc.

2-(2-Methoxyethoxy)phenylboronic acid

  • MDL number MFCD11183165
  • Cas Number : 1122568-09-5
  • Molecular Weight : 196.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1953
Availability details for
Quantity
  • Availability
  • Add to Cart