COM448618159 Combi-Blocks, Inc.

2-Allyloxyphenylboronic acid

  • MDL number MFCD11183169
  • Cas Number : 151414-76-5
  • Molecular Weight : 178.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1955
Availability details for
Quantity
  • Availability
  • Add to Cart