COM448618167 Combi-Blocks, Inc.

3,5-Diaminophenylboronic acid

  • MDL number MFCD11519193
  • Cas Number : 89641-16-7
  • Molecular Weight : 152.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2021
Availability details for
Quantity
  • Availability
  • Add to Cart