COM448618176 Combi-Blocks, Inc.

3,4-Bis(trifluoromethyl) phenylboronic acid

  • MDL number MFCD11617946
  • Cas Number : 1204745-88-9
  • Molecular Weight : 257.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-bis(trifluoromethyl)phenylboronic acid
Inchi Code 1S/C8H5BF6O2/c10-7(11,12)5-2-1-4(9(16)17)3-6(5)8(13,14)15/h1-3,16-17H
Inchi Key GIZQTQOXIWPFOA-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2121
Availability details for
Quantity
  • Availability
  • Add to Cart